Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Procyanidin A2 | 41743-41-3 | 5 MG
SDP

Catalog No. 5000420719
Encompass_Preferred

Procyanidin A2 is a flavonoid found in grapes, exhibiting anti-cancer, antioxidant, antimicrobial, and anti-inflammation activities. It is intended for research use only and not for sale to patients.

  • Purity: 99.81%
  • Molecular weight: 576.50
  • Formula: C30H24O12
  • Appearance: Solid
  • Color: White to pink
  • Structure classification: Flavonoids, biflavones, phenols, polyphenols
  • Initial source: Plants (Vitaceae, Vitis vinifera cv. Zalema, other families)
  • Solubility (in vitro): DMSO: 100 mg/mL (173.46 mM) (requires ultrasonic, hygroscopic DMSO has a significant impact on solubility)
  • Biological activity: Bacterial inhibitor, shows DPPH radical scavenging activity (IC50: 5.08 ± 0.37 μM), cytotoxicity against various human cell lines, and antiviral activity against HIV infected in human MT4 cells (IC50: 5.8 μg/mL).

Catalog No. 50-004-20719 Supplier Medchemexpress LLC Supplier No. HYN23435MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Procyanidin A2 is a flavonoid found in grapes, exhibiting anti-cancer, antioxidant, antimicrobial, and anti-inflammation activities. It is intended for research use only and not for sale to patients.

  • Purity: 99.81%
  • Molecular weight: 576.50
  • Formula: C30H24O12
  • Appearance: Solid
  • Color: White to pink
  • Structure classification: Flavonoids, biflavones, phenols, polyphenols
  • Initial source: Plants (Vitaceae, Vitis vinifera cv. Zalema, other families)
  • Solubility (in vitro): DMSO: 100 mg/mL (173.46 mM) (requires ultrasonic, hygroscopic DMSO has a significant impact on solubility)
  • Biological activity: Bacterial inhibitor, shows DPPH radical scavenging activity (IC50: 5.08 ± 0.37 μM), cytotoxicity against various human cell lines, and antiviral activity against HIV infected in human MT4 cells (IC50: 5.8 μg/mL).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.