Learn More
Medchemexpress LLC Procyanidin A2 | 41743-41-3 | MFCD10566019 | 99.8% | 576.50 g/mol | C30H24O12 | 10 MG
SDP
Catalog No. 5000289369
Encompass_Preferred
Procyanidin A2 is a naturally occurring flavonoid dimer isolated from grapes and other plant sources, provided as a research-grade analytical reagent for laboratory and analytical applications. It is used as an analytical standard and in biochemical studies exploring antioxidant, antimicrobial, anti-inflammatory, and anti-cancer effects.
  • High purity analytical standard suitable for research and analytical applications.
  • Reported biological activities include antioxidant, antimicrobial, anti-inflammatory, and anti-cancer effects.
  • Suitable for use in in vitro assays and analytical methods such as LC/MS.
  • Well-characterized chemical identity: CAS 41743-41-3, formula C30H24O12, molecular weight 576.50 g/mol.
  • Available in milligram-scale pack sizes for analytical workflows.
Catalog No. 50-002-89369 Supplier Medchemexpress LLC Supplier No. HYN234310MG
Procyanidin A2 is a naturally occurring flavonoid dimer isolated from grapes and other plant sources, provided as a research-grade analytical reagent for laboratory and analytical applications. It is used as an analytical standard and in biochemical studies exploring antioxidant, antimicrobial, anti-inflammatory, and anti-cancer effects.
  • High purity analytical standard suitable for research and analytical applications.
  • Reported biological activities include antioxidant, antimicrobial, anti-inflammatory, and anti-cancer effects.
  • Suitable for use in in vitro assays and analytical methods such as LC/MS.
  • Well-characterized chemical identity: CAS 41743-41-3, formula C30H24O12, molecular weight 576.50 g/mol.
  • Available in milligram-scale pack sizes for analytical workflows.