Learn More
AdipoGen Procyanidin A2
SDP
Catalog No. 502562511
Encompass_Preferred
Chemical. CAS 41743-41-3. Formula C30H24O12. MW 576.5. Isolated from grape seeds Vitis vinifera L. A-Type proanthocyanidine. Potent antioxidant. Free radical scavenger and lipid peroxidation inhibitor. Potent low-density lipoprotein LDL oxidation inhibitor. Antidiabetic. Prevents islet cell apoptosis, hyperglycemia and type 2 diabetes through its antioxidant properties. Antibacterial agent. Antiviral. Shown to inhibit viral replication of canine distemper virus CDV. Glycosyltransferase B GTFB inhibitor.
Catalog No. 50-256-2511 Supplier AdipoGen Supplier No. AGCN20486M005
Chemical. CAS 41743-41-3. Formula C30H24O12. MW 576.5. Isolated from grape seeds Vitis vinifera L. A-Type proanthocyanidine. Potent antioxidant. Free radical scavenger and lipid peroxidation inhibitor. Potent low-density lipoprotein LDL oxidation inhibitor. Antidiabetic. Prevents islet cell apoptosis, hyperglycemia and type 2 diabetes through its antioxidant properties. Antibacterial agent. Antiviral. Shown to inhibit viral replication of canine distemper virus CDV. Glycosyltransferase B GTFB inhibitor.