Learn More
Indofine Chemical Procyanidin B1, 1 Mg
SDP
Supplier:  Indofine Chemical 020983 1 MG
Encompass_Preferred
Molecular Formula: C30H26O12, Purity: 80% by HPLC, Molecular Weight: 578.53, Appearance: Powder, Storage conditions: Store at 0-8C, Also known as Epicatechin(4b-8)catechin
Catalog No. 50-909-059