Learn More
Medchemexpress LLC Procyanidin B1 | 20315-25-7 | 578.52 | 1 ML
SDP
Catalog No. 5000416718
Encompass_Preferred
Procyanidin B1 is a polyphenolic flavonoid isolated from commonly eaten fruits. It binds to the TLR4/MD-2 complex and exhibits anti-inflammatory activity.
  • Binds to TLR4/MD-2 complex
  • Possesses anti-inflammatory activity
  • Inhibits LPS-induced TNF-α production
  • Protects against Aβ oligomer-induced neuronal death
  • Induces expression of ACO and CPT1
Catalog No. 50-004-16718 Supplier Medchemexpress LLC Supplier No. HYN079510MM/1ML
Procyanidin B1 is a polyphenolic flavonoid isolated from commonly eaten fruits. It binds to the TLR4/MD-2 complex and exhibits anti-inflammatory activity.
  • Binds to TLR4/MD-2 complex
  • Possesses anti-inflammatory activity
  • Inhibits LPS-induced TNF-α production
  • Protects against Aβ oligomer-induced neuronal death
  • Induces expression of ACO and CPT1