Learn More
TARGETMOL CHEMICALS INC PROCYANIDIN B1 5MG
SDP
Catalog No. 502578813
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Procyanidin B1 isolated from commonly eaten fruits binds to the TLR4/MD-2 complex and has anti-inflammatory activity. purity: 99%
Catalog No. 50-257-8813 Supplier TARGETMOL CHEMICALS INC Supplier No. TQ02055MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Procyanidin B1 isolated from commonly eaten fruits binds to the TLR4/MD-2 complex and has anti-inflammatory activity. purity: 99%