Learn More
Medchemexpress LLC HY-N0795 5mg Medchemexpress, Procyanidin B1 CAS:20315-25-7 Purity:>98%
SDP
Catalog No. 501933721
Encompass_Preferred
Medchemexpress, HY-N0795 5mg Procyanidin B1 CAS:20315-25-7 Procyanidin B1 is a polyphenolic flavonoid isolated from commonly eaten fruits, binds to TLR4/MD-2 complex, and has anti-inflammatory activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3721 Supplier Medchemexpress LLC Supplier No. HYN07955MG
Medchemexpress, HY-N0795 5mg Procyanidin B1 CAS:20315-25-7 Procyanidin B1 is a polyphenolic flavonoid isolated from commonly eaten fruits, binds to TLR4/MD-2 complex, and has anti-inflammatory activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.