Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Procyanidin B1, ≥90% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111013464 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3464 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-3464 Supplier MilliporeSigma Supelco Supplier No. 195421MGF
Add to Cart
Add to Cart

Procyanidin B1 is a phenolic compound in fruits. It has been used widely as adulteration indicator of fruit juice. It has a strong antioxidant property.

Food and Beverages
TRUSTED_SUSTAINABILITY

Specifications

Packaging Poly Bottle
CAS 20315-25-7
Physical Form Neat
Linear Formula C30H26O12
MDL Number MFCD01861512
Synonym (-)-Epicatechin (4 beta-8)-(+)-catechin; cis,trans′′-4,8 ′′-Bi-(3,3 ′,4 ′,5,7-Pentahydroxyflavane); Proanthocyanidin B1
Type Analytical Standard
Quantity 1 mg
Formula Weight 578.52
Percent Purity ≥90% (HPLC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.