Learn More
Selleck Chemical LLC Procyanidin B1-E0240-5MG
SDP
Catalog No. 503137942
Encompass_Preferred
Procyanidin B1 a natural polyphenolic compound that has antioxidant properties inhibits Kv10 1 channel and suppresses the evolution of hepatoma
Catalog No. 50-313-7942 Supplier Selleck Chemical LLC Supplier No. E02405MG
Procyanidin B1 a natural polyphenolic compound that has antioxidant properties inhibits Kv10 1 channel and suppresses the evolution of hepatoma