Learn More
Medchemexpress LLC Procyanidin B2 | 29106-49-8 | 578.52 | 25 MG
SDP
Catalog No. 5000389243
Encompass_Preferred
Procyanidin B2 is a natural flavonoid known for its anti-cancer and antioxidant properties, making it valuable for various research applications. It is intended for research use only.
  • Exhibits antiproliferative activity in MCF-7 cells.
  • Inhibits NLRP3 inflammasome activation in human umbilical vein ECs (HUVECs).
  • Reduces reactive oxygen species (ROS).
  • Protects against cerebral ischemia-induced infarct volume and brain edema.
  • Improves neurological functional outcomes and regulates blood-brain barrier permeability.
  • Attenuates cerebral ischemia-induced tight junction degradation, mitochondrial depolarization, and intracellular oxidative stress.
  • Increases Nrf2 activation and expression of HO-1, GSTα, and NQO1 proteins.
Catalog No. 50-003-89243 Supplier Medchemexpress LLC Supplier No. 25MGHYN0796R
Procyanidin B2 is a natural flavonoid known for its anti-cancer and antioxidant properties, making it valuable for various research applications. It is intended for research use only.
  • Exhibits antiproliferative activity in MCF-7 cells.
  • Inhibits NLRP3 inflammasome activation in human umbilical vein ECs (HUVECs).
  • Reduces reactive oxygen species (ROS).
  • Protects against cerebral ischemia-induced infarct volume and brain edema.
  • Improves neurological functional outcomes and regulates blood-brain barrier permeability.
  • Attenuates cerebral ischemia-induced tight junction degradation, mitochondrial depolarization, and intracellular oxidative stress.
  • Increases Nrf2 activation and expression of HO-1, GSTα, and NQO1 proteins.