Learn More
Medchemexpress LLC Procyanidin B2 | 29106-49-8 | 578.52 | 50 MG
SDP
Catalog No. 5000416681
Encompass_Preferred
Procyanidin B2 is a natural flavonoid with established anti-cancer and antioxidant properties. This compound has been studied for its diverse biological activities, including its effects on cellular proliferation and its protective actions in ischemic conditions.
  • Inhibits NLRP3 inflammasome activation in human umbilical vein ECs (HUVECs).
  • Reduces reactive oxygen species (ROS) in HUVECs.
  • Demonstrates antiproliferative activity in MCF-7 cells.
  • Protects against cerebral ischemia-induced infarct volume and brain edema.
  • Improves functional outcomes and regulates blood-brain barrier permeability.
  • Attenuates tight junction degradation, mitochondrial depolarization, and intracellular oxidative stress.
  • Increases Nrf2 activation and the expression of HO-1, GSTα, and NQO1 proteins.
Catalog No. 50-004-16681 Supplier Medchemexpress LLC Supplier No. HYN0796R50MG
Procyanidin B2 is a natural flavonoid with established anti-cancer and antioxidant properties. This compound has been studied for its diverse biological activities, including its effects on cellular proliferation and its protective actions in ischemic conditions.
  • Inhibits NLRP3 inflammasome activation in human umbilical vein ECs (HUVECs).
  • Reduces reactive oxygen species (ROS) in HUVECs.
  • Demonstrates antiproliferative activity in MCF-7 cells.
  • Protects against cerebral ischemia-induced infarct volume and brain edema.
  • Improves functional outcomes and regulates blood-brain barrier permeability.
  • Attenuates tight junction degradation, mitochondrial depolarization, and intracellular oxidative stress.
  • Increases Nrf2 activation and the expression of HO-1, GSTα, and NQO1 proteins.