Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Procyanidin B2 | 29106-49-8 | MFCD01861513 | 99.9% | 578.52 | C30H26O12 | 10 MG
SDP

Catalog No. 5000296631
Encompass_Preferred

Procyanidin B2 analytical standard intended for research and analytical applications. Provided as a high-purity solid reference material for qualitative and quantitative method development and calibration in chromatographic and mass spectrometric assays. Ideal for laboratory standards, method validation, and instrument calibration.

  • High purity (~99.9%) suitable for analytical work.
  • Molecular weight 578.52 g/mol.
  • CAS number 29106-49-8 for unambiguous identification.
  • Appearance: solid, light yellow to light brown.
  • Suitable for HPLC, GC, and MS calibration and method development.
  • Store and handle according to the certificate of analysis and SDS.

Catalog No. 50-002-96631 Supplier Medchemexpress LLC Supplier No. HYN0796R10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Procyanidin B2 analytical standard intended for research and analytical applications. Provided as a high-purity solid reference material for qualitative and quantitative method development and calibration in chromatographic and mass spectrometric assays. Ideal for laboratory standards, method validation, and instrument calibration.

  • High purity (~99.9%) suitable for analytical work.
  • Molecular weight 578.52 g/mol.
  • CAS number 29106-49-8 for unambiguous identification.
  • Appearance: solid, light yellow to light brown.
  • Suitable for HPLC, GC, and MS calibration and method development.
  • Store and handle according to the certificate of analysis and SDS.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.