Learn More
Medchemexpress LLC Procyanidin B2 (Standard) | 29106-49-8 | 578.52 | 100 MG
SDP
Catalog No. 5000416932
Encompass_Preferred
Procyanidin B2 is an analytical standard and natural flavonoid with anti-cancer and antioxidant activities, intended for research and analytical applications. It is commonly utilized in qualitative, quantitative, and methodological research experiments.
  • Analytical standard for Procyanidin B2
  • Used in qualitative, quantitative, and methodological research experiments
  • Applicable in HPLC, GC, and MS
  • Possesses anti-cancer activities
  • Possesses antioxidant activities
Catalog No. 50-004-16932 Supplier Medchemexpress LLC Supplier No. HYN0796R100MG
Procyanidin B2 is an analytical standard and natural flavonoid with anti-cancer and antioxidant activities, intended for research and analytical applications. It is commonly utilized in qualitative, quantitative, and methodological research experiments.
  • Analytical standard for Procyanidin B2
  • Used in qualitative, quantitative, and methodological research experiments
  • Applicable in HPLC, GC, and MS
  • Possesses anti-cancer activities
  • Possesses antioxidant activities