Learn More
Selleck Chemical LLC Procyanidin C1-E0478-5MG
SDP
Catalog No. 502390001
Encompass_Preferred
Procyanidin C1 is a polyphenolic component of grape seed extract increases the healthspan and lifespan of mice through its action on senescent cells PCC1 is a natural senotherapeutic agent with in vivo activity and a potential intervention to delay alleviate or prevent age-related pathologies
Catalog No. 50-239-0001 Supplier Selleck Chemical LLC Supplier No. E04785MG
Procyanidin C1 is a polyphenolic component of grape seed extract increases the healthspan and lifespan of mice through its action on senescent cells PCC1 is a natural senotherapeutic agent with in vivo activity and a potential intervention to delay alleviate or prevent age-related pathologies