Learn More
Medchemexpress LLC Procyanidin C1 (PCC1) | 37064-30-5 | 5 MG
SDP
Catalog No. 5000421531
Encompass_Preferred
Procyanidin C1 (PCC1) is a natural polyphenol with oral activity. It causes DNA damage, cell cycle arrest, and induces apoptosis. It decreases Bcl-2 levels while enhancing BAX, caspase 3, and caspase 9 expression in cancer cells. Procyanidin C1 also exhibits senotherapeutic activity and extends the lifespan in mice.
  • Natural polyphenol
  • Oral activity
  • Causes DNA damage
  • Induces cell cycle arrest
  • Induces apoptosis by decreasing Bcl-2 and enhancing BAX, caspase 3, and 9 expression in cancer cells
  • Shows senotherapeutic activity
  • Increases lifespan in mice
Catalog No. 50-004-21531 Supplier Medchemexpress LLC Supplier No. HYN23425MG
Procyanidin C1 (PCC1) is a natural polyphenol with oral activity. It causes DNA damage, cell cycle arrest, and induces apoptosis. It decreases Bcl-2 levels while enhancing BAX, caspase 3, and caspase 9 expression in cancer cells. Procyanidin C1 also exhibits senotherapeutic activity and extends the lifespan in mice.
  • Natural polyphenol
  • Oral activity
  • Causes DNA damage
  • Induces cell cycle arrest
  • Induces apoptosis by decreasing Bcl-2 and enhancing BAX, caspase 3, and 9 expression in cancer cells
  • Shows senotherapeutic activity
  • Increases lifespan in mice