Learn More
Alkali Scientific ProCycle™ 0.1mL 96 Well Rigid™ PCR Plate, Full Skirt, Polycarbonate Frame, 10/Pack, Green Frame, Clear Well
SDP
Catalog No. 5000663102
Encompass_Preferred
ProCycle™ 0.1mL 96 Well Rigid™ PCR Plate, Full Skirt, Polycarbonate Frame, 10/Pack, Green Frame, Clear Well. This PCR plate combines strength, durability, and precision, making it perfect for high-throughput PCR applications. The full skirt design ensures compatibility with automated systems, while the green polycarbonate frame provides added structural support. Clear wells allow for easy sample observation.
Catalog No. 50-006-63102 Supplier Alkali Scientific Supplier No. PCRFS96G
Add to Cart
Add to Cart
ProCycle™ 0.1mL 96 Well Rigid™ PCR Plate, Full Skirt, Polycarbonate Frame, 10/Pack, Green Frame, Clear Well. This PCR plate combines strength, durability, and precision, making it perfect for high-throughput PCR applications. The full skirt design ensures compatibility with automated systems, while the green polycarbonate frame provides added structural support. Clear wells allow for easy sample observation.