Learn More
Alkali Scientific ProCycle™ 0.1mL 96 Well Rigid™ PCR Plate, Full Skirt, Polycarbonate Frame, 10/Pack, Red Frame, Clear Well
SDP
Catalog No. 5000662807
Encompass_Preferred
ProCycle™ 0.1mL 96 Well Rigid™ PCR Plate, Full Skirt, Polycarbonate Frame, 10/Pack, Red Frame, Clear Well. Featuring a full skirt design for automation compatibility, this PCR plate is essential for high-throughput PCR applications. The red polycarbonate frame ensures durability and strength, while the clear wells offer excellent sample visibility.
Catalog No. 50-006-62807 Supplier Alkali Scientific Supplier No. PCRFS96R
Add to Cart
Add to Cart
ProCycle™ 0.1mL 96 Well Rigid™ PCR Plate, Full Skirt, Polycarbonate Frame, 10/Pack, Red Frame, Clear Well. Featuring a full skirt design for automation compatibility, this PCR plate is essential for high-throughput PCR applications. The red polycarbonate frame ensures durability and strength, while the clear wells offer excellent sample visibility.