Learn More
Alkali Scientific ProCycle 0.2mL 8-strip PCR tub
SDP
Catalog No. 503028298
Encompass_Preferred
ProCycle0 2mL 8-strip PCR tubes with separate dome caps assorted colors 125x8 1000/PK with ultrathin sidewalls and bottoms for more uniform efficient temperature transfer and separate caps This PCR reaction strip is available with either separate needle pierceable flatcaps or dome-topped caps While easily opened and closed with one hand their positive sealing will fully protect the contents from evaporation during the whole thermal cycle The cap has an integral seal preventing contamination with surface of lid While more easily handled than single tubes the strip will precisely fit standard well spacing and can also be used with 8-channel hand-held pipettors They are manufactured under the most stringent conditions to attain the highest quality standards in the industry and customers have a choice of colorless or four non-cytotoxic non-metallic colors They are packaged in tamperproof resealable bags 0 2mL strip tubes with separate flat or domed caps Ultra-thin walls Positive sea
Catalog No. 50-302-8298 Supplier Alkali Scientific Supplier No. PC7063A
ProCycle0 2mL 8-strip PCR tubes with separate dome caps assorted colors 125x8 1000/PK with ultrathin sidewalls and bottoms for more uniform efficient temperature transfer and separate caps This PCR reaction strip is available with either separate needle pierceable flatcaps or dome-topped caps While easily opened and closed with one hand their positive sealing will fully protect the contents from evaporation during the whole thermal cycle The cap has an integral seal preventing contamination with surface of lid While more easily handled than single tubes the strip will precisely fit standard well spacing and can also be used with 8-channel hand-held pipettors They are manufactured under the most stringent conditions to attain the highest quality standards in the industry and customers have a choice of colorless or four non-cytotoxic non-metallic colors They are packaged in tamperproof resealable bags 0 2mL strip tubes with separate flat or domed caps Ultra-thin walls Positive sea