Learn More
Alkali Scientific ProCycle 0.2ml 96 Well PCR Pla
SDP
Supplier:  Alkali Scientific PCN8210
Encompass_Preferred
Introducing our ProCycle0 2ml 96 Well PCR Plate Semi Skirt White 10/pk 100/cs designed to elevate your PCR experiments with a host of exceptional features Versatile Thermal Cycler Compatibility Compatible with a wide range of thermal cyclers Effortless Well Identification Featuring high-contrast black alphanumeric indexing for easy well recognition Optimal Heat Transfer The ultra-thin uniform well design ensures maximum heat transfer enhancing reaction efficiency Minimized Sample Evaporation Well ridges help reduce sample evaporation when using film foil or strip caps for sealing Premium Materials Crafted from high-clarity ultra-pure polypropylene meeting FDA 21 CFR 177 1520 and USP Class VI standards Ideal for Various Industries Suitable for applications in the pharmaceutical food and molecular biology industries Quality Assurance Certified free of detectable DNase RNase DNA PCR inhibitors and rigorously tested for pyrogen-free performance Our PCR Plates are manufactured with h
Catalog No. 50-302-7662