Learn More
Alkali Scientific ProCycle™ 0.2mL 96 Well Rigid™ PCR Plate, Semi Skirted, Polycarbonate Frame, 10/Pack, Assorted Frame, Clear Well
SDP
Catalog No. 5000662762
Encompass_Preferred
ProCycle™ 0.2mL 96 Well Rigid™ PCR Plate, Semi Skirted, Polycarbonate Frame, 10/Pack, Assorted Frame, Clear Well. Offering versatility in a variety of research settings, this PCR plate comes in assorted colors and is designed for high-throughput PCR applications. Its semi-skirted design ensures compatibility with automated systems, and the clear wells provide enhanced sample visibility.
Catalog No. 50-006-62762 Supplier Alkali Scientific Supplier No. PCRS96AST
Add to Cart
Add to Cart
ProCycle™ 0.2mL 96 Well Rigid™ PCR Plate, Semi Skirted, Polycarbonate Frame, 10/Pack, Assorted Frame, Clear Well. Offering versatility in a variety of research settings, this PCR plate comes in assorted colors and is designed for high-throughput PCR applications. Its semi-skirted design ensures compatibility with automated systems, and the clear wells provide enhanced sample visibility.