Learn More
Alkali Scientific ProCycle™ 0.2mL 96 Well Rigid™ PCR Plate, Semi Skirted, Polycarbonate Frame, 10/Pack, Purple Frame, Clear Well
SDP
Catalog No. 5000662922
Encompass_Preferred
ProCycle™ 0.2mL 96 Well Rigid™ PCR Plate, Semi Skirted, Polycarbonate Frame, 10/Pack, Purple Frame, Clear Well. Designed for high-performance PCR workflows, this PCR plate offers a robust polycarbonate frame in purple for added durability. The semi-skirted design ensures excellent compatibility with most laboratory automation equipment, and the clear wells allow for precise sample observation and handling.
Catalog No. 50-006-62922 Supplier Alkali Scientific Supplier No. PCRS96V
Add to Cart
Add to Cart
ProCycle™ 0.2mL 96 Well Rigid™ PCR Plate, Semi Skirted, Polycarbonate Frame, 10/Pack, Purple Frame, Clear Well. Designed for high-performance PCR workflows, this PCR plate offers a robust polycarbonate frame in purple for added durability. The semi-skirted design ensures excellent compatibility with most laboratory automation equipment, and the clear wells allow for precise sample observation and handling.