Learn More
Alkali Scientific ProCycle 0.2mL CLEAR 8-Strip T
SDP
Catalog No. 503028299
Encompass_Preferred
ProCycle0 2mL CLEAR with Attached Hinged Flat Cap Strip eliminates the need to carry two separate components in inventory The strip includes 8 integral 0 2 mL tubes with ultra-thin sidewalls and bottoms for more uniform and efficient temperature transfer While easily opened and closed with one hand their positive sealing will fully protect the contents from evaporation during the whole thermal cycle The cap has an integral seal preventing contamination with surface of lid While more easily handled than single tubes the 8-strip PCR tubes will precisely fit standard well spacing and can also be used with multi-channel hand-held pipettors They are made of ultra-clear polypropylene plastic under the most stringent conditions to attain the highest quality standards in the industry and strips can be cut to any length 0 2mL 8-strip PCR tubes with attached caps Caps are Optically Clear for qPCR Ultra-thin walls for uniformity and consistency Positive sealing eliminates any evaporation F
Catalog No. 50-302-8299 Supplier Alkali Scientific Supplier No. PC7066F
ProCycle0 2mL CLEAR with Attached Hinged Flat Cap Strip eliminates the need to carry two separate components in inventory The strip includes 8 integral 0 2 mL tubes with ultra-thin sidewalls and bottoms for more uniform and efficient temperature transfer While easily opened and closed with one hand their positive sealing will fully protect the contents from evaporation during the whole thermal cycle The cap has an integral seal preventing contamination with surface of lid While more easily handled than single tubes the 8-strip PCR tubes will precisely fit standard well spacing and can also be used with multi-channel hand-held pipettors They are made of ultra-clear polypropylene plastic under the most stringent conditions to attain the highest quality standards in the industry and strips can be cut to any length 0 2mL 8-strip PCR tubes with attached caps Caps are Optically Clear for qPCR Ultra-thin walls for uniformity and consistency Positive sealing eliminates any evaporation F