Learn More
Alkali Scientific ProCycle 0.2mL StableStrip 8-S
SDP
Catalog No. 503029257
Encompass_Preferred
ProCycle StableStrip 0 2 mL 8-strip PCR tubes include individually attached hinged caps which are either needle pierceable flat caps or domed caps This eliminates the need to carry two separate components in inventory The strip includes 8 integral 0 2 mL tubes with ultra-thin sidewalls and bottoms for more uniform and efficient temperature transfer While easily opened and closed with one hand their positive sealing will fully protect the contents from evaporation during the whole thermal cycle The cap has an integral seal preventing contamination with surface of lid While more easily handled than single tubes the 8-strip PCR tubes will precisely fit standard well spacing and can also be used with multi-channel hand-held pipettors They are made of ultra-clear polypropylene plastic under the most stringent conditions to attain the highest quality standards in the industry and strips can be cut to any length 0 2mL 8-strip PCR tubes with attached caps Caps are Optically Clear for
Catalog No. 50-302-9257 Supplier Alkali Scientific Supplier No. PC7060A
ProCycle StableStrip 0 2 mL 8-strip PCR tubes include individually attached hinged caps which are either needle pierceable flat caps or domed caps This eliminates the need to carry two separate components in inventory The strip includes 8 integral 0 2 mL tubes with ultra-thin sidewalls and bottoms for more uniform and efficient temperature transfer While easily opened and closed with one hand their positive sealing will fully protect the contents from evaporation during the whole thermal cycle The cap has an integral seal preventing contamination with surface of lid While more easily handled than single tubes the 8-strip PCR tubes will precisely fit standard well spacing and can also be used with multi-channel hand-held pipettors They are made of ultra-clear polypropylene plastic under the most stringent conditions to attain the highest quality standards in the industry and strips can be cut to any length 0 2mL 8-strip PCR tubes with attached caps Caps are Optically Clear for