Learn More
Alkali Scientific ProCycle™ 0.5mL Thin-Walled Individual Tubes with Frosted Flat Caps, Sterile, 1000/Pk
SDP
Catalog No. 503029108
Encompass_Preferred
ProCycle™ 0.5mL Thin-Walled Individual Tubes with flat caps ProCycle 0.2mL Individual PCR Tubes are designed for oil-free operation, and are made of transparent, superior quality grade polypropylene for better viewing of the contents. Their ultra-thin wall design ensures rapid thermal transfer and a significant reduction in cycle and PCR reaction time. Attached flat-topped hinged caps can be used with heated thermal cycler lids. The caps provide positive sealing during thermal cycling and will prevent evaporation while being easily opened and closed with one hand. The cap has an integral shield which prevents contamination with the lid surface, and has frosted writing surface for sample identification. They are packaged in tamperproof, resealable safety-lock bags. Rnase/Dnase, DNA & Pyrogen Free Polypropylene plastic High clarity Frosted flat cap Positive sealing to prevent evaporation Reduction in cycle and PCR reaction times 1000 per pack
Catalog No. 50-302-9108 Supplier Alkali Scientific Supplier No. PC7069F
ProCycle™ 0.5mL Thin-Walled Individual Tubes with flat caps ProCycle 0.2mL Individual PCR Tubes are designed for oil-free operation, and are made of transparent, superior quality grade polypropylene for better viewing of the contents. Their ultra-thin wall design ensures rapid thermal transfer and a significant reduction in cycle and PCR reaction time. Attached flat-topped hinged caps can be used with heated thermal cycler lids. The caps provide positive sealing during thermal cycling and will prevent evaporation while being easily opened and closed with one hand. The cap has an integral shield which prevents contamination with the lid surface, and has frosted writing surface for sample identification. They are packaged in tamperproof, resealable safety-lock bags. Rnase/Dnase, DNA & Pyrogen Free Polypropylene plastic High clarity Frosted flat cap Positive sealing to prevent evaporation Reduction in cycle and PCR reaction times 1000 per pack