Learn More
Alkali Scientific ProCycle™ 0.1mL 8-Strip PCR Tubes, Low Profile with Attached, Clear, Flat Caps 120X8, 960/cs
SDP
Catalog No. 502335749
Encompass_Preferred
This more convenient 0.1 ml low-profile tube strip comes with caps. The strip includes 8 integral 0.1 ml tubes with ultra-thin sidewalls and bottoms for more uniform and efficient temperature transfer. This PCR reaction strip is available with flat (needle pierceable) caps. While easily opened and closed with one hand, their positive sealing will fully protect the contents from evaporation during the whole thermal cycle. The cap has an integral seal preventing contamination with surface of lid. While more easily handled than single tubes, the strip will precisely fit standard well spacing and can also be used with 8-channel hand-held pipettors. Manufactured under the most stringent conditions to attain the highest quality standards in the industry. Choice of colorless and four non-cytotoxic and non-metallic colors. Packed in tamperproof resealable bags. 960/pk
Catalog No. 50-233-5749 Supplier Alkali Scientific Supplier No. PC7065
This more convenient 0.1 ml low-profile tube strip comes with caps. The strip includes 8 integral 0.1 ml tubes with ultra-thin sidewalls and bottoms for more uniform and efficient temperature transfer. This PCR reaction strip is available with flat (needle pierceable) caps. While easily opened and closed with one hand, their positive sealing will fully protect the contents from evaporation during the whole thermal cycle. The cap has an integral seal preventing contamination with surface of lid. While more easily handled than single tubes, the strip will precisely fit standard well spacing and can also be used with 8-channel hand-held pipettors. Manufactured under the most stringent conditions to attain the highest quality standards in the industry. Choice of colorless and four non-cytotoxic and non-metallic colors. Packed in tamperproof resealable bags. 960/pk