Learn More
Alkali Scientific ProCycle™ 0.2mL 8-Strip PCR Tubes with Separate Flat Strip Caps 125 X 8, 1000/cs
SDP
Catalog No. 502335966
Encompass_Preferred
The 0.2mL Rigid Stable 8-strip PCR tube incorporates individually attached flat caps for more convenience. No need to carry two separate components in inventory. The strip includes 8 integral 0.2mL tubes with ultra-thin sidewalls and bottoms for more uniform and efficient temperature transfer. This PCR reaction strip is available with seperate flat (needle pierceable) caps. Their positive sealing will fully protect the contents from evaporation during the whole thermal cycle. The cap has an integral seal preventing contamination with surface of the lid. While more easily handled than single tubes, the 8-strip PCR tubes will precisely fit standard well spacing and can also be used with multi-channel hand-held pipettors. Manufactured under the most stringent conditions to attain the highest quality standards in the industry. Tubes are made of ultra-clear polypropylene plastic and strips can be cut to any length. 1000/pk
Catalog No. 50-233-5966 Supplier Alkali Scientific Supplier No. PC7064
The 0.2mL Rigid Stable 8-strip PCR tube incorporates individually attached flat caps for more convenience. No need to carry two separate components in inventory. The strip includes 8 integral 0.2mL tubes with ultra-thin sidewalls and bottoms for more uniform and efficient temperature transfer. This PCR reaction strip is available with seperate flat (needle pierceable) caps. Their positive sealing will fully protect the contents from evaporation during the whole thermal cycle. The cap has an integral seal preventing contamination with surface of the lid. While more easily handled than single tubes, the 8-strip PCR tubes will precisely fit standard well spacing and can also be used with multi-channel hand-held pipettors. Manufactured under the most stringent conditions to attain the highest quality standards in the industry. Tubes are made of ultra-clear polypropylene plastic and strips can be cut to any length. 1000/pk