Learn More
Alkali Scientific ProCycle 0.2mL 8-Strip PCR tubes with Separate Dome Strip Caps, 125x8, 1000/pk
SDP
Catalog No. 502336118
Encompass_Preferred
Alkali Scientific’s ProCycleª 0.2 mL 8-strip PCR tubes include eight integral 0.2 mL tubes with ultrathin sidewalls and bottoms for more uniform, efficient temperature transfer and separate caps. This PCR reaction strip is available with either separate needle pierceable flatcaps or dome-topped caps. While easily opened and closed with one hand, their positive sealing will fully protect the contents from evaporation during the whole thermal cycle. The cap has an integral seal preventing contamination with surface of lid. While more easily handled than single tubes, the strip will precisely fit standard well spacing and can also be used with 8-channel hand-held pipettors. They are manufactured under the most stringent conditions to attain the highest quality standards in the industry, and customers have a choice of colorless or four non-cytotoxic, non-metallic colors. They are packaged in tamperproof resealable bags.
Catalog No. 50-233-6118 Supplier Alkali Scientific Supplier No. PC7063
Alkali Scientific’s ProCycleª 0.2 mL 8-strip PCR tubes include eight integral 0.2 mL tubes with ultrathin sidewalls and bottoms for more uniform, efficient temperature transfer and separate caps. This PCR reaction strip is available with either separate needle pierceable flatcaps or dome-topped caps. While easily opened and closed with one hand, their positive sealing will fully protect the contents from evaporation during the whole thermal cycle. The cap has an integral seal preventing contamination with surface of lid. While more easily handled than single tubes, the strip will precisely fit standard well spacing and can also be used with 8-channel hand-held pipettors. They are manufactured under the most stringent conditions to attain the highest quality standards in the industry, and customers have a choice of colorless or four non-cytotoxic, non-metallic colors. They are packaged in tamperproof resealable bags.