Learn More
Alkali Scientific ProCycle™ 0.2mL Rigid stable 8-Strip PCR Tubes with Attached Individual Flat Caps, ASSORTED COLORS, 120x8, 960/PK
SDP
Catalog No. 502335905
Encompass_Preferred
The StableStrip™ 0.2 ml 8-strip PCR tube incorporates individually attached caps (flat or domed) for more convenience. No need to carry two separate components in inventory. The strip includes 8 integral 0.2 ml tubes with ultra-thin sidewalls and bottoms for more uniform and efficient temperature transfer. This PCR reaction strip is available with either flat (needle pierceable) or dome-topped individually attached hinged caps. While easily opened and closed with one hand, their positive sealing will fully protect the contents from evaporation during the whole thermal cycle. The cap has an integral seal preventing contamination with surface of lid.While more easily handled than single tubes, the 8-strip PCR tubes will precisely fit standard well spacing and can also be used with multi-channel hand-held pipetters. Manufactured under the most stringent conditions to attain the highest quality standards in the industry.Tubes are made of ultra-clear polypropylene plastic and
Catalog No. 50-233-5905 Supplier Alkali Scientific Supplier No. PC7061A
The StableStrip™ 0.2 ml 8-strip PCR tube incorporates individually attached caps (flat or domed) for more convenience. No need to carry two separate components in inventory. The strip includes 8 integral 0.2 ml tubes with ultra-thin sidewalls and bottoms for more uniform and efficient temperature transfer. This PCR reaction strip is available with either flat (needle pierceable) or dome-topped individually attached hinged caps. While easily opened and closed with one hand, their positive sealing will fully protect the contents from evaporation during the whole thermal cycle. The cap has an integral seal preventing contamination with surface of lid.While more easily handled than single tubes, the 8-strip PCR tubes will precisely fit standard well spacing and can also be used with multi-channel hand-held pipetters. Manufactured under the most stringent conditions to attain the highest quality standards in the industry.Tubes are made of ultra-clear polypropylene plastic and