Learn More
Alkali Scientific ProCycle™ 0.2mL StableStrip™ 8-Strip PCR Tubes with Attached Individual Dome Caps, 120 X 8, 960/cs
SDP
Catalog No. 502336051
Encompass_Preferred
The StableStrip™ 0.2 mL 8-strip PCR tube incorporates individually attached dome caps for more convenience. No need to carry two separate components in inventory. The strip includes 8 integral 0.2 mL tubes with ultra-thin sidewalls and bottoms for more uniform and efficient temperature transfer. This PCR reaction strip is available with dome-topped individually attached hinged caps. While easily opened and closed with one hand, their positive sealing will fully protect the contents from evaporation during the whole thermal cycle. The cap has an integral seal preventing contamination with surface of the lid. While more easily handled than single tubes, these 8-strip PCR tubes will precisely fit standard well spacing and can also be used with multi-channel hand-held pipettors. Manufactured under the most stringent conditions to attain the highest quality standards in the industry. Tubes are made of ultra-clear polypropylene plastic and strips can be cut to any length. 960/pk
Catalog No. 50-233-6051 Supplier Alkali Scientific Supplier No. PC7060
The StableStrip™ 0.2 mL 8-strip PCR tube incorporates individually attached dome caps for more convenience. No need to carry two separate components in inventory. The strip includes 8 integral 0.2 mL tubes with ultra-thin sidewalls and bottoms for more uniform and efficient temperature transfer. This PCR reaction strip is available with dome-topped individually attached hinged caps. While easily opened and closed with one hand, their positive sealing will fully protect the contents from evaporation during the whole thermal cycle. The cap has an integral seal preventing contamination with surface of the lid. While more easily handled than single tubes, these 8-strip PCR tubes will precisely fit standard well spacing and can also be used with multi-channel hand-held pipettors. Manufactured under the most stringent conditions to attain the highest quality standards in the industry. Tubes are made of ultra-clear polypropylene plastic and strips can be cut to any length. 960/pk