Learn More
Alkali Scientific ProCycle™ 1.2 mL 8-Strip Cluster (Dilution) Tubes, Racked, 125/Pk, Sterile
SDP
Catalog No. 503027281
Encompass_Preferred
ProCycle™ 1.2 mL 8-Strip Cluster Tubes, Racked Systems are ideal for high-throughput screening processes and long-term storage. It all begins with high-clarity, ultra-pure polypropylene, combined with proprietary injection molding techniques to produce high-clarity vessels that are exceptionally robust, chemically-resistant, and pure. SBS footprint; 8 × 12 format. Ideal for automation / HTS. Tubes and racks are made from virgin polypropylene. Tube-and-rack system is suitable as a reaction vessel in your robotic workstation or as a long-term storage vessel in your sample library. A clear lid is a standard part of racked system. Certified free of detectable RNase DNase, DNA, PCR inhibitors, and tested pyrogen-free.
Catalog No. 50-302-7281 Supplier Alkali Scientific Supplier No. CTR8012S
ProCycle™ 1.2 mL 8-Strip Cluster Tubes, Racked Systems are ideal for high-throughput screening processes and long-term storage. It all begins with high-clarity, ultra-pure polypropylene, combined with proprietary injection molding techniques to produce high-clarity vessels that are exceptionally robust, chemically-resistant, and pure. SBS footprint; 8 × 12 format. Ideal for automation / HTS. Tubes and racks are made from virgin polypropylene. Tube-and-rack system is suitable as a reaction vessel in your robotic workstation or as a long-term storage vessel in your sample library. A clear lid is a standard part of racked system. Certified free of detectable RNase DNase, DNA, PCR inhibitors, and tested pyrogen-free.