Learn More
Alkali Scientific ProCycle™ 1.2 mL Individual Cluster (Dilution) Tubes, Racked, 1000/Pk, Non-Sterile
SDP
Catalog No. 503027113
Encompass_Preferred
ProCycle™ 1.2 mL Individual Cluster Tubes, Racked are ideal for high-throughput screening processes and long-term storage. It all begins with high-clarity, ultra-pure polypropylene, combined with proprietary injection molding techniques to produce high-clarity vessels that are exceptionally robust, chemically-resistant, and pure. SBS footprint; 8 × 12 format. Ideal for automation / HTS. Tubes and racks are made from virgin polypropylene Tube-and-rack system is suitable as a reaction vessel in your robotic workstation or as a long-term storage vessel in your sample library. A clear lid is a standard part of racked system. Certified free of detectable RNase DNase, DNA, PCR inhibitors, and tested pyrogen-free.
Catalog No. 50-302-7113 Supplier Alkali Scientific Supplier No. CTR1012
ProCycle™ 1.2 mL Individual Cluster Tubes, Racked are ideal for high-throughput screening processes and long-term storage. It all begins with high-clarity, ultra-pure polypropylene, combined with proprietary injection molding techniques to produce high-clarity vessels that are exceptionally robust, chemically-resistant, and pure. SBS footprint; 8 × 12 format. Ideal for automation / HTS. Tubes and racks are made from virgin polypropylene Tube-and-rack system is suitable as a reaction vessel in your robotic workstation or as a long-term storage vessel in your sample library. A clear lid is a standard part of racked system. Certified free of detectable RNase DNase, DNA, PCR inhibitors, and tested pyrogen-free.