Learn More
Alkali Scientific ProCycle™ 30µL 384 Well Rigid™ PCR Plate, Full Skirt, A24 Notch, Polycarbonate Frame, 10/Pack, Blue Frame, Clear Well
SDP
Catalog No. 5000663017
Encompass_Preferred
ProCycle™ 30µL 384 Well Rigid™ PCR Plate, Full Skirt, A24 Notch, Polycarbonate Frame, 10/Pack, Blue Frame, Clear Well. This blue-framed PCR plate provides a clear view of the sample wells and is designed for rigorous PCR amplification processes. The full skirt ensures stability, and the polycarbonate frame offers extra support for precise pipetting.
Catalog No. 50-006-63017 Supplier Alkali Scientific Supplier No. PCR384B
Add to Cart
Add to Cart
ProCycle™ 30µL 384 Well Rigid™ PCR Plate, Full Skirt, A24 Notch, Polycarbonate Frame, 10/Pack, Blue Frame, Clear Well. This blue-framed PCR plate provides a clear view of the sample wells and is designed for rigorous PCR amplification processes. The full skirt ensures stability, and the polycarbonate frame offers extra support for precise pipetting.