Learn More
Alkali Scientific ProCycle™ 30µL 384 Well Rigid™ PCR Plate, Full Skirt, A24 Notch, Polycarbonate Frame, 10/Pack, Red Frame, Clear Well
SDP
Catalog No. 5000662117
Encompass_Preferred
ProCycle™ 30µL 384 Well Rigid™ PCR Plate, Full Skirt, A24 Notch, Polycarbonate Frame, 10/Pack, Red Frame, Clear Well. This red-framed PCR plate is specifically designed to enhance sample handling in PCR workflows. Its full skirt design ensures a stable foundation during processing, while the polycarbonate frame offers strength and clarity for precise sample visualization.
Catalog No. 50-006-62117 Supplier Alkali Scientific Supplier No. PCR384R
Add to Cart
Add to Cart
ProCycle™ 30µL 384 Well Rigid™ PCR Plate, Full Skirt, A24 Notch, Polycarbonate Frame, 10/Pack, Red Frame, Clear Well. This red-framed PCR plate is specifically designed to enhance sample handling in PCR workflows. Its full skirt design ensures a stable foundation during processing, while the polycarbonate frame offers strength and clarity for precise sample visualization.