Learn More
Alkali Scientific ProCycle™ 30µL 384 Well Rigid™ PCR Plate, Full Skirt, A24 & P24 Notches, Polycarbonate Frame, 10/Pack, Clear Frame, White Well
SDP
Catalog No. 5000662951
Encompass_Preferred
ProCycle™ 30µL 384 Well Rigid™ PCR Plate, Full Skirt, A24 & P24 Notches, Polycarbonate Frame, 10/Pack, Clear Frame, White Well. Designed for efficient PCR applications, this rigid PCR plate offers excellent stability and durability with a clear polycarbonate frame and white wells to enhance sample visibility. Ideal for use in molecular biology laboratories.
Catalog No. 50-006-62951 Supplier Alkali Scientific Supplier No. PCR384IIW
Add to Cart
Add to Cart
ProCycle™ 30µL 384 Well Rigid™ PCR Plate, Full Skirt, A24 & P24 Notches, Polycarbonate Frame, 10/Pack, Clear Frame, White Well. Designed for efficient PCR applications, this rigid PCR plate offers excellent stability and durability with a clear polycarbonate frame and white wells to enhance sample visibility. Ideal for use in molecular biology laboratories.