Learn More
Alkali Scientific ProCycle™ 384 Well PCR Plates, Full Skirt Edge, A24 Notch, 10/cs
SDP
Catalog No. 502336174
Encompass_Preferred
This PCR plate is compatible with a broad range of thermal cyclers, ensuring versatility and adaptability in your lab. It offers easy well identification with high-contrast black alphanumeric indexing that facilitates quick and accurate well identification, streamlining your workflow. The ultra-thin, uniform wells ensure optimal heat transfer, promoting high reaction efficiency and reproducibility. The well ridges help reduce sample evaporation when sealing with film, foil, or strip caps, preserving the integrity of your samples. This PCR plate is manufactured from high-clarity, ultra-pure polypropylene, meeting FDA 21 CFR 177.1520 and USP Class VI standards. PC384 offers purity assurance and is certified free of detectable DNase, RNase, DNA, PCR inhibitors, and has been tested to be pyrogen-free.
Catalog No. 50-233-6174 Supplier Alkali Scientific Supplier No. PC384
This PCR plate is compatible with a broad range of thermal cyclers, ensuring versatility and adaptability in your lab. It offers easy well identification with high-contrast black alphanumeric indexing that facilitates quick and accurate well identification, streamlining your workflow. The ultra-thin, uniform wells ensure optimal heat transfer, promoting high reaction efficiency and reproducibility. The well ridges help reduce sample evaporation when sealing with film, foil, or strip caps, preserving the integrity of your samples. This PCR plate is manufactured from high-clarity, ultra-pure polypropylene, meeting FDA 21 CFR 177.1520 and USP Class VI standards. PC384 offers purity assurance and is certified free of detectable DNase, RNase, DNA, PCR inhibitors, and has been tested to be pyrogen-free.