Learn More
Alkali Scientific ProCycle™ Clear Roche 480 LightCycler, qPCR Plate, Full Edge, 384 Well, 10/Pk
SDP
Catalog No. 503029086
Encompass_Preferred
Broad thermal cycler compatibility. High-contrast, black alphanumeric indexing for easy well identification. Ultra-thin, uniform wells ensure optimal heat transfer for high reaction efficiency. Well ridges reduce sample evaporation when sealing with film, foil, or strip caps. Manufactured from high-clarity, ultra-pure polypropylene (meets FDA 21 CFR 177.1520 & USP Class VI). For use in pharmaceutical industry, food industry, and molecular biology. Certified free of detectable DNase, RNase, DNA, PCR inhibitors, and tested pyrogen-free. Roche LightCycler 480 is a registered trademark of F. Hoffmann-La Roche Ltd, Roche Applied Science
Catalog No. 50-302-9086 Supplier Alkali Scientific Supplier No. PC3RC
Broad thermal cycler compatibility. High-contrast, black alphanumeric indexing for easy well identification. Ultra-thin, uniform wells ensure optimal heat transfer for high reaction efficiency. Well ridges reduce sample evaporation when sealing with film, foil, or strip caps. Manufactured from high-clarity, ultra-pure polypropylene (meets FDA 21 CFR 177.1520 & USP Class VI). For use in pharmaceutical industry, food industry, and molecular biology. Certified free of detectable DNase, RNase, DNA, PCR inhibitors, and tested pyrogen-free. Roche LightCycler 480 is a registered trademark of F. Hoffmann-La Roche Ltd, Roche Applied Science