Learn More
Alkali Scientific ProCycle™ Fast Optical qPCR Plates, Half-Skirted, 96X0.1mL Well, ABi Style A1notch, 10/cs
SDP
Catalog No. 502335975
Encompass_Preferred
PCLP96 feature exceptional thermal conductivity for precise thermal cycling. Consistently delivers reproducible, specific, and sensitive PCR results in just 25 minutes. Validated with other system components to ensure consistent outcomes. Compatible with the following machines: Life Technologies/ThermoFisher StepOne Plus, Life Technologies/ThermoFisher Quantstudio 1, Life Technologies/ThermoFisher Quantstudio 3, Life Technologies/ThermoFisher Quantstudio 5, Life Technologies/ThermoFisher , Quantstudio 6 Pro and 7, and Life Technologies/ThermoFisher Quantstudio 12K FLEX **most popular**. 10/pk
Catalog No. 50-233-5975 Supplier Alkali Scientific Supplier No. PCLP96
PCLP96 feature exceptional thermal conductivity for precise thermal cycling. Consistently delivers reproducible, specific, and sensitive PCR results in just 25 minutes. Validated with other system components to ensure consistent outcomes. Compatible with the following machines: Life Technologies/ThermoFisher StepOne Plus, Life Technologies/ThermoFisher Quantstudio 1, Life Technologies/ThermoFisher Quantstudio 3, Life Technologies/ThermoFisher Quantstudio 5, Life Technologies/ThermoFisher , Quantstudio 6 Pro and 7, and Life Technologies/ThermoFisher Quantstudio 12K FLEX **most popular**. 10/pk