Learn More
Alkali Scientific ProCycle™ Flat Optical 8-Cap Strips for qPCR/PCR Tubes & Plates, 125/Pk
SDP
Catalog No. 503028243
Encompass_Preferred
For use in pharmaceutical industry, food industry, and molecular biology. Manufactured from high-quality, virgin polypropylene to ensure purity and provide excellent optical clarity. Certified free of detectable RNase, DNase, DNA, PCR inhibitors, and tested pyrogen-free.
Catalog No. 50-302-8243 Supplier Alkali Scientific Supplier No. PC7041
For use in pharmaceutical industry, food industry, and molecular biology. Manufactured from high-quality, virgin polypropylene to ensure purity and provide excellent optical clarity. Certified free of detectable RNase, DNase, DNA, PCR inhibitors, and tested pyrogen-free.