Learn More
Alkali Scientific ProCycle™ Full Skirted PCR Plates, Full Edge, Clear, 96 x 0.1mL Well, Printed Alphanumerics, 10/Pk
SDP
Catalog No. 502336144
Encompass_Preferred
Universal Compatibility: This PCR plate is compatible with a broad range of thermal cyclers, ensuring versatility and adaptability in your lab. Easy Well Identification: High-contrast, black alphanumeric indexing facilitates quick and accurate well identification, streamlining your workflow. Optimal Heat Transfer: Ultra-thin, uniform wells ensure optimal heat transfer, promoting high reaction efficiency and reproducibility. Sample Protection: Well ridges help reduce sample evaporation when sealing with film, foil, or strip caps, preserving the integrity of your samples. High Clarity, Ultra-Pure Material: This PCR plate is manufactured from high-clarity, ultra-pure polypropylene, meeting FDA 21 CFR 177.1520 and USP Class VI standards. Purity Assurance: The plate is certified free of detectable DNase, RNase, DNA, PCR inhibitors, and has been tested to be pyrogen-free.
Catalog No. 50-233-6144 Supplier Alkali Scientific Supplier No. PCFS96
Universal Compatibility: This PCR plate is compatible with a broad range of thermal cyclers, ensuring versatility and adaptability in your lab. Easy Well Identification: High-contrast, black alphanumeric indexing facilitates quick and accurate well identification, streamlining your workflow. Optimal Heat Transfer: Ultra-thin, uniform wells ensure optimal heat transfer, promoting high reaction efficiency and reproducibility. Sample Protection: Well ridges help reduce sample evaporation when sealing with film, foil, or strip caps, preserving the integrity of your samples. High Clarity, Ultra-Pure Material: This PCR plate is manufactured from high-clarity, ultra-pure polypropylene, meeting FDA 21 CFR 177.1520 and USP Class VI standards. Purity Assurance: The plate is certified free of detectable DNase, RNase, DNA, PCR inhibitors, and has been tested to be pyrogen-free.