Learn More
Alkali Scientific ProCycle Full Skirted PCR Plat
SDP
Catalog No. 503028948
Encompass_Preferred
Introducing our ProCycleFull Skirted PCR Plates Full Edge 96 x 0 1mL Well (Clear) designed to elevate your PCR experiments with a host of exceptional features Universal Compatibility This PCR plate is compatible with a broad range of thermal cyclers ensuring versatility and adaptability in your lab Easy Well Identification High-contrast black alphanumeric indexing facilitates quick and accurate well identification streamlining your workflow Optimal Heat Transfer Ultra-thin uniform wells ensure optimal heat transfer promoting high reaction efficiency and reproducibility Sample Protection Well ridges help reduce sample evaporation when sealing with film foil or strip caps preserving the integrity of your samples High Clarity Ultra-Pure Material This PCR plate is manufactured from high-clarity ultra-pure polypropylene meeting FDA 21 CFR 177 1520 and USP Class VI standards Purity Assurance The plate is certified free of detectable DNase RNase DNA PCR inhibitors and has been tested to
Catalog No. 50-302-8948 Supplier Alkali Scientific Supplier No. PCFS96W
Introducing our ProCycleFull Skirted PCR Plates Full Edge 96 x 0 1mL Well (Clear) designed to elevate your PCR experiments with a host of exceptional features Universal Compatibility This PCR plate is compatible with a broad range of thermal cyclers ensuring versatility and adaptability in your lab Easy Well Identification High-contrast black alphanumeric indexing facilitates quick and accurate well identification streamlining your workflow Optimal Heat Transfer Ultra-thin uniform wells ensure optimal heat transfer promoting high reaction efficiency and reproducibility Sample Protection Well ridges help reduce sample evaporation when sealing with film foil or strip caps preserving the integrity of your samples High Clarity Ultra-Pure Material This PCR plate is manufactured from high-clarity ultra-pure polypropylene meeting FDA 21 CFR 177 1520 and USP Class VI standards Purity Assurance The plate is certified free of detectable DNase RNase DNA PCR inhibitors and has been tested to