Learn More
Alkali Scientific ProCycle™ Mini Skirted PCR Plates, Half-Skirted, 96 x 0.2ml Well, BLACK PRINTED Alphanumerics, A12 cut corner, 10/PK
SDP
Catalog No. 502335906
Encompass_Preferred
PCS96 Compatible with the following machines:• ABI 7000• ABI 7300• ABI 7500• ABI 7700• ABI StepOne• BioRad iCycler• BioRad myIQ• BioRad myIQ5• ABI Sequencer 3100• ABI Sequencer 3132
Catalog No. 50-233-5906 Supplier Alkali Scientific Supplier No. PCS96
PCS96 Compatible with the following machines:• ABI 7000• ABI 7300• ABI 7500• ABI 7700• ABI StepOne• BioRad iCycler• BioRad myIQ• BioRad myIQ5• ABI Sequencer 3100• ABI Sequencer 3132