Learn More
Alkali Scientific ProCycle™ PCR Tubes, Individual 0.2mL Flat Cap, 1000/cs
SDP
Catalog No. 502336123
Encompass_Preferred
Designed for oil-free operation, these tubes are made of transparent superior quality grade polypropylene for better viewing of the contents. Their ultra-thin wall design will ensure rapid thermal transfer and a significant reduction in cycle and PCR reaction time. Attached hinged caps are flat-topped and can be used with heated thermal cycler lids. They provide positive sealing during thermal cycling and will prevent evaporation while being easily opened and closed with one hand. The cap has an integral shield preventing contamination with surface of lid. Frosted writing surface for sample identification. Packaged in tamperproof resealable safety-lock bags. 1000/pk
Catalog No. 50-233-6123 Supplier Alkali Scientific Supplier No. PC7067F
Designed for oil-free operation, these tubes are made of transparent superior quality grade polypropylene for better viewing of the contents. Their ultra-thin wall design will ensure rapid thermal transfer and a significant reduction in cycle and PCR reaction time. Attached hinged caps are flat-topped and can be used with heated thermal cycler lids. They provide positive sealing during thermal cycling and will prevent evaporation while being easily opened and closed with one hand. The cap has an integral shield preventing contamination with surface of lid. Frosted writing surface for sample identification. Packaged in tamperproof resealable safety-lock bags. 1000/pk