Learn More
Alkali Scientific ProCycle™ PCR Tubes, Individual 0.2m with Dome Cap (1000/Pk)
SDP
Catalog No. 503028991
Encompass_Preferred
ProCycle™ PCR Tubes, Individual 0.2mL domed cap (1000/Pk), and are made of transparent, superior quality grade polypropylene for better viewing of the contents. Their ultra-thin wall design ensures rapid thermal transfer and a significant reduction in cycle and PCR reaction time. Attached hinged caps are either dome or flat-topped, and can be used with heated thermal cycler lids. The caps provide positive sealing during thermal cycling and will prevent evaporation while being easily opened and closed with one hand. The cap has an integral shield which prevents contamination with the lid surface, and has frosted writing surface for sample identification. They are packaged in tamperproof, resealable safety-lock bags. Rnase/Dnase, DNA & Pyrogen Free Polypropylene plastic High clarity Flat or domed caps Positive sealing to prevent evaporation Reduction in cycle and PCR reaction times 1000 per pack
Catalog No. 50-302-8991 Supplier Alkali Scientific Supplier No. PC7068D
ProCycle™ PCR Tubes, Individual 0.2mL domed cap (1000/Pk), and are made of transparent, superior quality grade polypropylene for better viewing of the contents. Their ultra-thin wall design ensures rapid thermal transfer and a significant reduction in cycle and PCR reaction time. Attached hinged caps are either dome or flat-topped, and can be used with heated thermal cycler lids. The caps provide positive sealing during thermal cycling and will prevent evaporation while being easily opened and closed with one hand. The cap has an integral shield which prevents contamination with the lid surface, and has frosted writing surface for sample identification. They are packaged in tamperproof, resealable safety-lock bags. Rnase/Dnase, DNA & Pyrogen Free Polypropylene plastic High clarity Flat or domed caps Positive sealing to prevent evaporation Reduction in cycle and PCR reaction times 1000 per pack