Learn More
Alkali Scientific ProCycle™ Pre-cut Sealing Membranes, Sterile, Aluminum membrane, 10 bags of 10 membranes
SDP
Catalog No. 503028653
Encompass_Preferred
RNase, DNase, DNA, and PCR inhibitor free Evaporation rates ≤ 5% Inner packs of 10 membranes PureAmp™ Sealing Membranes are compatible with all standard PCR plates. Membrane options include pierceable aluminum, standard transparent, optically clear sticky adhesive, optically clear pressure-sensitive adhesive, and low-adhesive EZ-peel, all of which are guaranteed to keep evaporation rates below 5%.
Catalog No. 50-302-8653 Supplier Alkali Scientific Supplier No. SF1001AS
RNase, DNase, DNA, and PCR inhibitor free Evaporation rates ≤ 5% Inner packs of 10 membranes PureAmp™ Sealing Membranes are compatible with all standard PCR plates. Membrane options include pierceable aluminum, standard transparent, optically clear sticky adhesive, optically clear pressure-sensitive adhesive, and low-adhesive EZ-peel, all of which are guaranteed to keep evaporation rates below 5%.