Learn More
Alkali Scientific ProCycle™ Sep-R-8™ Breakable PCR Plates, Standard 96 well x 0.2ml, non-skirted, breakable into strips of 8, 50/pk
SDP
Catalog No. 503028329
Encompass_Preferred
Optimize your PCR experiments with the Sep-R 8 Breakable PCR Plates. Designed for versatility and convenience, these non-skirted, 96-well plates can be easily broken into strips of 8, allowing for flexible sample management and reduced waste. With a capacity of 0.2ml per well, these plates are ideal for high-throughput applications, ensuring reliable performance and reproducibility in your PCR workflows. Product Specifications: Plate Format: Standard 96-well Well Volume: 0.2ml per well Skirting: Non-skirted Breakability: Breakable into strips of 8 Material: High-quality, low-binding plastic for minimal sample loss Compatibility: Suitable for most PCR and qPCR thermocyclers Pack Size: 50 plates per pack Enhance your laboratory efficiency with Sep-R 8 Breakable PCR Plates, providing the flexibility and precision required for your molecular biology applications.
Catalog No. 50-302-8329 Supplier Alkali Scientific Supplier No. PC9602
Optimize your PCR experiments with the Sep-R 8 Breakable PCR Plates. Designed for versatility and convenience, these non-skirted, 96-well plates can be easily broken into strips of 8, allowing for flexible sample management and reduced waste. With a capacity of 0.2ml per well, these plates are ideal for high-throughput applications, ensuring reliable performance and reproducibility in your PCR workflows. Product Specifications: Plate Format: Standard 96-well Well Volume: 0.2ml per well Skirting: Non-skirted Breakability: Breakable into strips of 8 Material: High-quality, low-binding plastic for minimal sample loss Compatibility: Suitable for most PCR and qPCR thermocyclers Pack Size: 50 plates per pack Enhance your laboratory efficiency with Sep-R 8 Breakable PCR Plates, providing the flexibility and precision required for your molecular biology applications.