Learn More
Alkali Scientific ProCycle Standard PCR Plates W
SDP
Catalog No. 503028330
Encompass_Preferred
Introducing our ProCycleStandard PCR Plates Non-Skirted 96 x 0 2ml Well BLACK PRINTED Alphanumerics A12 cut corner 10/Pk designed to elevate your PCR experiments with a host of exceptional features Universal Compatibility This PCR plate is compatible with a broad range of thermal cyclers ensuring versatility and adaptability in your lab Easy Well Identification High-contrast black alphanumeric indexing facilitates quick and accurate well identification streamlining your workflow Optimal Heat Transfer Ultra-thin uniform wells ensure optimal heat transfer promoting high reaction efficiency and reproducibility Sample Protection Well ridges help reduce sample evaporation when sealing with film foil or strip caps preserving the integrity of your samples High Clarity Ultra-Pure Material This PCR plate is manufactured from high-clarity ultra-pure polypropylene meeting FDA 21 CFR 177 1520 and USP Class VI standards Purity Assurance The plate is certified free of detectable DNase RNase DNA
Catalog No. 50-302-8330 Supplier Alkali Scientific Supplier No. PCF96W
Introducing our ProCycleStandard PCR Plates Non-Skirted 96 x 0 2ml Well BLACK PRINTED Alphanumerics A12 cut corner 10/Pk designed to elevate your PCR experiments with a host of exceptional features Universal Compatibility This PCR plate is compatible with a broad range of thermal cyclers ensuring versatility and adaptability in your lab Easy Well Identification High-contrast black alphanumeric indexing facilitates quick and accurate well identification streamlining your workflow Optimal Heat Transfer Ultra-thin uniform wells ensure optimal heat transfer promoting high reaction efficiency and reproducibility Sample Protection Well ridges help reduce sample evaporation when sealing with film foil or strip caps preserving the integrity of your samples High Clarity Ultra-Pure Material This PCR plate is manufactured from high-clarity ultra-pure polypropylene meeting FDA 21 CFR 177 1520 and USP Class VI standards Purity Assurance The plate is certified free of detectable DNase RNase DNA