Learn More
Alkali Scientific ProCycle™ Standard PCR Plates, Non-Skirted, 96 x 0.2ml Well, BLACK PRINTED Alphanumerics, A12 cut corner, 10/Pk
SDP
Catalog No. 502335967
Encompass_Preferred
PCF96 Compatible with the following machines:• MJ/BioRad Opticon• Opticon 2• MiniOpticon• Chromo4• Agilent/Stratagene MX4000P• MX3000P• MX3000i• MX3005PPCS96 Compatible with the following machines:• ABI 7000• ABI 7300• ABI 7500• ABI 7700• ABI StepOne• BioRad iCycler• BioRad myIQ• BioRad myIQ5• ABI Sequencer 3100• ABI Sequencer 3130
Catalog No. 50-233-5967 Supplier Alkali Scientific Supplier No. PCF96
PCF96 Compatible with the following machines:• MJ/BioRad Opticon• Opticon 2• MiniOpticon• Chromo4• Agilent/Stratagene MX4000P• MX3000P• MX3000i• MX3005PPCS96 Compatible with the following machines:• ABI 7000• ABI 7300• ABI 7500• ABI 7700• ABI StepOne• BioRad iCycler• BioRad myIQ• BioRad myIQ5• ABI Sequencer 3100• ABI Sequencer 3130