Learn More
Alkali Scientific ProCycle™ Two-Component PCR Plate, 96-Well, Semi-Skirted PC Frame, 0.25mL White PP Wells, 50/pk
SDP
Catalog No. 503028304
Encompass_Preferred
SureFrame™ plates are made with a special two-material process that creates an ultra-rigid polycarbonate frame while maintaining the favorable thermal transfer characteristics of thin-walled polypropylene tube wells. This ultra-rigid frame is ideal for all automated platforms and plate grippers and increases the uniformity of your experiments. Traditional single-material plates have been shown to warp inside thermal cyclers. This deformation of the plates causes increased evaporation in the outer wells of the plate, leading to inconsistent amplification across wells. SureFrame’s polycarbonate frame eliminates warping, for completely uniform results.   Two material design for increased rigidity Ideal for automated platforms Reduces evaporation Certified for PCR
Catalog No. 50-302-8304 Supplier Alkali Scientific Supplier No. PCR9096HSW
SureFrame™ plates are made with a special two-material process that creates an ultra-rigid polycarbonate frame while maintaining the favorable thermal transfer characteristics of thin-walled polypropylene tube wells. This ultra-rigid frame is ideal for all automated platforms and plate grippers and increases the uniformity of your experiments. Traditional single-material plates have been shown to warp inside thermal cyclers. This deformation of the plates causes increased evaporation in the outer wells of the plate, leading to inconsistent amplification across wells. SureFrame’s polycarbonate frame eliminates warping, for completely uniform results.   Two material design for increased rigidity Ideal for automated platforms Reduces evaporation Certified for PCR