Learn More
Alkali Scientific ProCycle White 0.1mL Low-Profi
SDP
Catalog No. 503028295
Encompass_Preferred
ProCycle0 1mL 8-strip PCR tubes low profile with attached clear flat caps 120x8 960/PK This more convenient 0 1 ml low-profile tube strip comes with caps The strip includes 8 integral 0 1 ml tubes with ultra-thin sidewalls and bottoms for more uniform and efficient temperature transfer This PCR reaction strip is available with flat (needle pierceable) caps While easily opened and closed with one hand their positive sealing will fully protect the contents from evaporation during the whole thermal cycle The cap has an integral seal preventing contamination with surface of lid While more easily handled than single tubes the strip will precisely fit standard well spacing and can also be used with 8-channel hand-held pipettors Manufactured under the most stringent conditions to attain the highest quality standards in the industry Choice of colorless and four non-cytotoxic and non-metallic colors Packed in tamperproof resealable bags White tubes offer enhanced fluorescence reflectio
Catalog No. 50-302-8295 Supplier Alkali Scientific Supplier No. PC7065W
ProCycle0 1mL 8-strip PCR tubes low profile with attached clear flat caps 120x8 960/PK This more convenient 0 1 ml low-profile tube strip comes with caps The strip includes 8 integral 0 1 ml tubes with ultra-thin sidewalls and bottoms for more uniform and efficient temperature transfer This PCR reaction strip is available with flat (needle pierceable) caps While easily opened and closed with one hand their positive sealing will fully protect the contents from evaporation during the whole thermal cycle The cap has an integral seal preventing contamination with surface of lid While more easily handled than single tubes the strip will precisely fit standard well spacing and can also be used with 8-channel hand-held pipettors Manufactured under the most stringent conditions to attain the highest quality standards in the industry Choice of colorless and four non-cytotoxic and non-metallic colors Packed in tamperproof resealable bags White tubes offer enhanced fluorescence reflectio