Learn More
Alkali Scientific ProCycle™ White Fast Optical qPCR Plates, Half-Skirted, 96 x 0.1ml Well, ABi Style A1 notch, 10/Pk
SDP
Catalog No. 503028247
Encompass_Preferred
The Alkali Scientiic™ Procycle™ White Fast Optical 96-Well Reaction Plates are a direct replacement for your existing Applied Biosystems MicroAmp Fast Optical 96-Well Reaction Plates. Key Features: Exceptional thermal conductivity for precise thermal cycling. Consistently delivers reproducible, specific, and sensitive PCR results in just 25 minutes. Validated with other system components to ensure consistent outcomes. PCLP96 is compatible with the following machines: Life Technologies/ThermoFisher StepOne Plus Life Technologies/ThermoFisher Quantstudio 1 Life Technologies/ThermoFisher Quantstudio 3 Life Technologies/ThermoFisher Quantstudio 5 Life Technologies/ThermoFisher Quantstudio 6 Pro & 7 Life Technologies/ThermoFisher Quantstudio 12K FLEX (*most popular)
Catalog No. 50-302-8247 Supplier Alkali Scientific Supplier No. PCLP96W
The Alkali Scientiic™ Procycle™ White Fast Optical 96-Well Reaction Plates are a direct replacement for your existing Applied Biosystems MicroAmp Fast Optical 96-Well Reaction Plates. Key Features: Exceptional thermal conductivity for precise thermal cycling. Consistently delivers reproducible, specific, and sensitive PCR results in just 25 minutes. Validated with other system components to ensure consistent outcomes. PCLP96 is compatible with the following machines: Life Technologies/ThermoFisher StepOne Plus Life Technologies/ThermoFisher Quantstudio 1 Life Technologies/ThermoFisher Quantstudio 3 Life Technologies/ThermoFisher Quantstudio 5 Life Technologies/ThermoFisher Quantstudio 6 Pro & 7 Life Technologies/ThermoFisher Quantstudio 12K FLEX (*most popular)